Learn More
Enzo Life Sciences C2 Ceramide, (5mg), CAS Number: 3102-57-6
SDP
Supplier:  Enzo Life Sciences BMLSL1000005
Encompass_Preferred
Cell-permeable ceramide analog. N-Acetylsphingosine is a biologically active, cell-permeable ceramide analog. It inhibits cell proliferation and induces monocytic differentiation of HL-60 cells and induces apoptosis. It stimulates protein phosphatase 2A4, activates MAP kinase and SAP kinase and induces PKCδ and ε translocation. Physiological levels of C2 ceramide, produced by the action of a PAF:sphingosine CoA-independent transacetylase, have been detected in HL-60 cells. Alternative Name: N-Acetyl-D-erythro-sphingosine. Formula: C20H39NO3. MW: 341.5. CAS Number: 3102-57-6. Purity: ≥98% (TLC). Appearance: White solid. Solubility: Soluble in 100% ethanol (>25mg/ml) or DMSO (>50mg/ml). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC9629861
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.